Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP12
Entrez Gene ID
102116452
Gene Name
fatty acid binding protein 12
Gene Aliases
-
Chromosome Location
Chr.8: 82704618 - 82714055 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563622.2 XP_005563679.1 1 - 2 105 99
XM_015455000.1 XP_015310486.1 1 - 2 105 99

View Full Product Details