Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIB1
Entrez Gene ID
102121314
Gene Name
tribbles pseudokinase 1
Gene Aliases
-
Chromosome Location
Chr.8: 126811000 - 126819260 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564057.2 XP_005564114.1 2 - 3 1428 89
XM_015455168.1 XP_015310654.1 2 - 3 386 89
XM_015455169.1 XP_015310655.1 2 - 3 380 89

View Full Product Details