Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH4
Entrez Gene ID
102120275
Gene Name
junctophilin 4
Gene Aliases
-
Chromosome Location
Chr.7: 87287127 - 87298105 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560876.2 XP_005560933.1 5 - 6 2772 92
XM_005560877.2 XP_005560934.1 4 - 5 2646 92

View Full Product Details