Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB26
Entrez Gene ID
102120027
Gene Name
zinc finger and BTB domain containing 26
Gene Aliases
-
Chromosome Location
Chr.15: 13113616 - 13127224 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580830.2 XP_005580887.1 2 - 2 3335 69
XM_005580831.2 XP_005580888.1 2 - 2 1375 69
XM_005580832.2 XP_005580889.1 2 - 2 1331 69
XM_005580833.2 XP_005580890.1 2 - 2 2109 69

View Full Product Details