Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGA
Entrez Gene ID
102138376
Gene Name
glycoprotein hormones, alpha polypeptide
Gene Aliases
CG-alpha, FSH-alpha, LSH-alpha, TSH-alpha
Chromosome Location
Chr.4: 86807321 - 86824021 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284544.1 NP_001271473.1 1 - 2 118 99
XM_015448839.1 XP_015304325.1 2 - 3 403 99
GenBank mRNA AJ781394.1 - 1 - 2 118 99
AY026358.1 - 1 - 2 103 99

View Full Product Details