Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT22
Entrez Gene ID
102119946
Gene Name
intraflagellar transport 22
Gene Aliases
RABL5
Chromosome Location
Chr.3: 45644640 - 45652089 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283136.1 NP_001270065.1 2 - 3 200 98
GenBank mRNA AB169889.1 - 2 - 3 200 98

View Full Product Details