Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC185
Entrez Gene ID
102120790
Gene Name
coiled-coil domain containing 185
Gene Aliases
C1H1orf65
Chromosome Location
Chr.1: 82252408 - 82254502 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005540938.1 XP_005540995.1 1 - 1 1910 98

View Full Product Details