Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHD2
Entrez Gene ID
102129262
Gene Name
EF-hand domain family member D2
Gene Aliases
-
Chromosome Location
Chr.1: 213089712 - 213109489 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015442482.1 XP_015297968.1 2 - 3 414 80

View Full Product Details