Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP6
Entrez Gene ID
102142720
Gene Name
aquaporin 6
Gene Aliases
-
Chromosome Location
Chr.11: 48828112 - 48833374 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570796.2 XP_005570853.1 1 - 2 1575 65
XM_015430896.1 XP_015286382.1 1 - 2 1574 65
XM_015430897.1 XP_015286383.1 1 - 2 988 65

View Full Product Details