Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQP6
Entrez Gene ID
102142720
Gene Name
aquaporin 6
Gene Aliases
-
Chromosome Location
Chr.11: 48828112 - 48833374 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570796.2 XP_005570853.1 4 - 5 1890 67

View Full Product Details