Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYRP1
Entrez Gene ID
102120654
Gene Name
tyrosinase related protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 66066624 - 66084988 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015437168.1 XP_015292654.1 6 - 7 1501 86

View Full Product Details