Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARTN
Entrez Gene ID
102123590
Gene Name
artemin
Gene Aliases
-
Chromosome Location
Chr.1: 183635730 - 183641928 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015437122.1 XP_015292608.1 3 - 4 4249 76

View Full Product Details