Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXA1
Entrez Gene ID
102125775
Gene Name
forkhead box A1
Gene Aliases
-
Chromosome Location
Chr.7: 101587190 - 101592980 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561120.1 XP_005561177.1 1 - 2 271 106

View Full Product Details