Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSF5
Entrez Gene ID
102133994
Gene Name
heat shock transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.16: 43943937 - 44007148 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583833.2 XP_005583890.1 4 - 5 1792 65

View Full Product Details