Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KISS1
Entrez Gene ID
102122810
Gene Name
KiSS-1 metastasis suppressor
Gene Aliases
-
Chromosome Location
Chr.1: 62667251 - 62673971 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015448612.1 XP_015304098.1 2 - 3 555 61

View Full Product Details