Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Afp
Entrez Gene ID
11576
Gene Name
alpha fetoprotein
Gene Aliases
-
Chromosome Location
Chr.5: 90490714 - 90508907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007423.4 NP_031449.3 10 - 11 1345 109
GenBank mRNA AK010934.1 - 10 - 11 1321 109
AK075972.1 - 10 - 11 1325 109
AK076053.1 - 10 - 11 1321 109
AK076197.1 - 10 - 11 1323 109
AK145975.1 - 10 - 11 1321 109
AK146513.1 - 10 - 11 1323 109
AK146524.1 - 10 - 11 1323 109
AK167549.1 - 10 - 11 1321 109
AK168426.1 - 10 - 11 1324 109
AK168574.1 - 10 - 11 1323 109
AK168576.1 - 10 - 11 1321 109
AK168751.1 - 10 - 11 1323 109
AK168818.1 - 9 - 10 1199 109
AK168883.1 - 10 - 11 1321 109
AK169054.1 - 10 - 11 1323 109
AK169147.1 - 10 - 11 1321 109
AV105248.2 - 1 - 2 264 109
BC066206.1 - 10 - 11 1308 109
V00743.1 - 10 - 11 1319 109

View Full Product Details