Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bmp6
Entrez Gene ID
12161
Gene Name
bone morphogenetic protein 6
Gene Aliases
D13Wsu115e, Vgr1
Chromosome Location
Chr.13: 38345716 - 38499728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007556.2 NP_031582.1 6 - 7 1600 71
GenBank mRNA AK014797.1 - - 138 71
AK041210.1 - - 1958 71
AK135770.1 - 6 - 7 1233 71
BC138593.1 - 6 - 7 1520 71
BC138595.1 - 6 - 7 1520 71
J04566.1 - 6 - 7 1182 71
X80992.1 - 6 - 7 1599 71

View Full Product Details