The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008168.2 | NP_032194.2 | 11 - 12 | 1652 | 57 |
GenBank mRNA | BC052009.1 | - | 11 - 12 | 1540 | 57 |
BC058097.1 | - | 11 - 12 | 1411 | 57 | |
BC110682.1 | - | 11 - 12 | 1575 | 57 | |
D10011.1 | - | 11 - 12 | 1581 | 57 |