Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspa4
Entrez Gene ID
15525
Gene Name
heat shock protein 4
Gene Aliases
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025
Chromosome Location
Chr.11: 53259814 - 53300479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008300.3 NP_032326.3 15 - 16 2243 126
GenBank mRNA AK053601.1 - 15 - 16 2177 126
AK054211.1 - 8 - 9 965 126
AK133772.1 - 15 - 16 2186 126
AK148883.1 - 4 - 5 518 126
AK155305.1 - 15 - 16 2160 126
AK166102.1 - 15 - 16 2226 126
AK220167.1 - 15 - 16 2194 126
BC003950.1 - 15 - 16 2138 124
CA316420.1 - 4 - 5 538 126
D85904.1 - 15 - 16 1942 126

View Full Product Details