The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001254747.1 | NP_001241676.1 | 4 - 5 | 607 | 73 |
NM_008357.2 | NP_032383.1 | 4 - 5 | 617 | 73 | |
GenBank mRNA | AK085441.1 | - | 6 - 7 | 817 | 73 |
AK155616.1 | - | 4 - 5 | 1255 | 73 | |
BC023698.1 | - | 4 - 5 | 602 | 73 | |
CJ137432.1 | - | 1 - 2 | 73 | ||
DQ083236.1 | - | 1 - 2 | 111 | 73 | |
DQ083237.1 | - | 1 - 2 | 111 | 73 | |
U14332.1 | - | 4 - 5 | 576 | 73 |