Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Masp1
Entrez Gene ID
17174
Gene Name
mannan-binding lectin serine peptidase 1
Gene Aliases
AW048060, CCPII, Crarf, Masp1/3
Chromosome Location
Chr.16: 23451785 - 23521073 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008555.2 NP_032581.2 12 - 13 1606 79
GenBank mRNA BC131637.1 - 12 - 13 1606 79
BC131638.1 - 12 - 13 1606 79
D16492.1 - - 1619 79

View Full Product Details