Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptn
Entrez Gene ID
19242
Gene Name
pleiotrophin
Gene Aliases
HARP, HB-GAM, HBBN, HBGF-8, HBNF, OSF, Osf-1, Osf1
Chromosome Location
Chr.6: 36714909 - 36811361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008973.2 NP_032999.1 4 - 5 1921 75
GenBank mRNA AK011346.1 - 4 - 5 778 75
AK161964.1 - 4 - 5 612 75
BC002064.1 - 4 - 5 490 75
BC061695.1 - 4 - 5 721 75
D90225.1 - 4 - 5 665 75
EF437379.1 - 1 - 2 152 75

View Full Product Details