The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243072.1 | NP_001230001.1 | 13 - 14 | 1828 | 82 |
NM_001243073.1 | NP_001230002.1 | 13 - 14 | 1668 | 82 | |
NM_009152.4 | NP_033178.2 | 12 - 13 | 2096 | 82 | |
GenBank mRNA | AK140817.1 | - | 13 - 14 | 1669 | 82 |
BC057588.1 | - | 12 - 13 | 1743 | 82 | |
BC090844.1 | - | 13 - 14 | 1828 | 82 | |
D85028.2 | - | 12 - 13 | 2098 | 82 | |
L40484.1 | - | 9 - 10 | 1129 | 82 | |
L41541.1 | - | 12 - 13 | 1447 | 82 | |
X85993.1 | - | 12 - 13 | 1557 | 82 |