The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112699.1 | NP_001106169.1 | 6 - 7 | 574 | 87 |
NM_001112700.1 | NP_001106170.1 | 6 - 7 | 514 | 87 | |
NM_011180.3 | NP_035310.2 | 6 - 7 | 574 | 87 | |
GenBank mRNA | AB013464.1 | - | 6 - 7 | 495 | 87 |
AB035538.1 | - | 6 - 7 | 543 | 87 | |
AF051337.1 | - | 5 - 6 | 442 | 87 | |
AK078502.1 | - | - | 484 | 87 | |
AK158216.1 | - | 6 - 7 | 514 | 87 | |
AK172296.1 | - | 6 - 7 | 519 | 87 | |
AY126516.1 | - | 6 - 7 | 543 | 87 | |
BC057974.1 | - | 6 - 7 | 458 | 87 |