Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt2
Entrez Gene ID
21956
Gene Name
troponin T2, cardiac
Gene Aliases
Tnt, cTnT
Chromosome Location
Chr.1: 135836316 - 135852268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130174.2 NP_001123646.1 9 - 10 443 76
NM_001130175.2 NP_001123647.4 9 - 10 439 76
NM_001130176.1 NP_001123648.1 7 - 8 330 76
NM_001130177.2 NP_001123649.1 8 - 9 431 76
NM_001130178.2 NP_001123650.1 8 - 9 413 76
NM_001130179.2 NP_001123651.4 8 - 9 409 76
NM_001130180.2 NP_001123652.1 7 - 8 401 76
NM_001130181.2 NP_001123653.4 7 - 8 397 76
NM_011619.3 NP_035749.1 7 - 8 401 76
GenBank mRNA AK146945.1 - 9 - 10 388 76
AK163786.1 - 8 - 9 376 76
AK168393.1 - 8 - 9 376 76
AK169061.1 - 8 - 9 359 76
BC063753.1 - 9 - 10 419 76
BQ554639.1 - 8 - 9 349 76
L47549.1 - 8 - 9 327 76
L47550.1 - 8 - 9 312 76
L47551.1 - 7 - 8 313 76
L47552.1 - 7 - 8 322 76
L47553.1 - 7 - 8 329 76
L47599.1 - 8 - 9 333 76
L47600.1 - 7 - 8 330 76

View Full Product Details