Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt2
Entrez Gene ID
21956
Gene Name
troponin T2, cardiac
Gene Aliases
Tnt, cTnT
Chromosome Location
Chr.1: 135836316 - 135852268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130174.2 NP_001123646.1 15 - 16 991 67
NM_001130175.2 NP_001123647.4 15 - 16 987 67
NM_001130176.1 NP_001123648.1 14 - 15 887 67
NM_001130177.2 NP_001123649.1 14 - 15 979 67
NM_001130178.2 NP_001123650.1 15 - 16 970 67
NM_001130179.2 NP_001123651.4 15 - 16 966 67
NM_001130180.2 NP_001123652.1 14 - 15 958 67
NM_001130181.2 NP_001123653.4 14 - 15 954 67
NM_011619.3 NP_035749.1 13 - 14 949 67
GenBank mRNA AK146945.1 - 15 - 16 936 67
AK168393.1 - 14 - 15 924 67
AK169061.1 - 15 - 16 916 67
BC063753.1 - 15 - 16 967 67
L47549.1 - 15 - 16 884 67
L47550.1 - 15 - 16 869 67
L47551.1 - 14 - 15 870 67
L47552.1 - 14 - 15 879 67
L47553.1 - 14 - 15 886 67
L47599.1 - 15 - 16 890 67
L47600.1 - 13 - 14 878 67

View Full Product Details