Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syt8
Entrez Gene ID
55925
Gene Name
synaptotagmin VIII
Gene Aliases
-
Chromosome Location
Chr.7: 142434462 - 142440399 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285857.1 NP_001272786.1 7 - 8 922 126
NM_001285858.1 NP_001272787.1 7 - 8 933 126
NM_001285861.1 NP_001272790.1 7 - 8 923 126
NM_018802.5 NP_061272.2 7 - 8 1044 126
GenBank mRNA AB026805.1 - 7 - 8 923 126
AK075832.1 - 7 - 8 1045 126
BC100747.1 - 4 - 5 757 126
BC100748.1 - 7 - 8 898 126
BC103602.1 - 7 - 8 898 126
BC103665.1 - 7 - 8 909 126
U20107.1 - 5 - 6 704 126

View Full Product Details