The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019394.3 | NP_062267.2 | 2 - 3 | 361 | 59 |
GenBank mRNA | AW047492.1 | - | 2 - 3 | 59 | |
BE864464.1 | - | 2 - 3 | 361 | 59 | |
CK794645.1 | - | 2 - 3 | 355 | 59 | |
X94322.1 | - | 2 - 3 | 366 | 59 |