The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019431.2 | NP_062304.1 | 2 - 3 | 303 | 64 |
GenBank mRNA | AJ272045.1 | - | 2 - 3 | 303 | 64 |
AK144253.1 | - | 2 - 3 | 534 | 64 | |
BC138503.1 | - | 2 - 3 | 322 | 64 | |
BC138506.1 | - | 2 - 3 | 322 | 64 |