The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293748.1 | NP_001280677.2 | 1 - 2 | 199 | 65 |
NM_022314.3 | NP_071709.3 | 1 - 2 | 199 | 65 | |
GenBank mRNA | AF317223.1 | - | 1 - 2 | 113 | 65 |
BC029186.1 | - | 1 - 2 | 156 | 65 | |
BY770803.1 | - | 1 - 2 | 200 | 65 | |
U04541.1 | - | 1 - 2 | 169 | 65 |