The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023580.4 | NP_076069.2 | 9 - 10 | 1774 | 63 |
GenBank mRNA | AF131197.1 | - | 9 - 10 | 1716 | 63 |
AK028478.1 | - | 9 - 10 | 1775 | 63 | |
BC071215.1 | - | 9 - 10 | 1738 | 63 | |
U18084.1 | - | 5 - 6 | 877 | 63 |