Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Epha1
Entrez Gene ID
13835
Gene Name
Eph receptor A1
Gene Aliases
5730453L17Rik, AL033318, Eph, Esk
Chromosome Location
Chr.6: 42358487 - 42373310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023580.4 NP_076069.2 13 - 14 2206 113
GenBank mRNA AF131197.1 - 13 - 14 2148 113
AK028478.1 - 13 - 14 2207 113
BC071215.1 - 13 - 14 2170 113

View Full Product Details