Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfxn2
Entrez Gene ID
94279
Gene Name
sideroflexin 2
Gene Aliases
C79732, F630107H02Rik
Chromosome Location
Chr.19: 46573322 - 46596901 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053196.3 NP_444426.3 3 - 4 443 64
GenBank mRNA AF325261.1 - 3 - 4 401 64
AK034875.1 - - 443 64
AK149277.1 - - 412 64
AK150722.1 - 3 - 4 394 64
AK162407.1 - 3 - 4 443 64
BC019808.1 - 3 - 4 441 64

View Full Product Details