Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pim3
Entrez Gene ID
223775
Gene Name
proviral integration site 3
Gene Aliases
BC026639, Kid1
Chromosome Location
Chr.15: 88862194 - 88865726 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145478.2 NP_663453.1 4 - 5 1093 75
GenBank mRNA AK149478.1 - 4 - 5 813 75
AK159399.1 - 4 - 5 812 75
AK159600.1 - 4 - 5 812 75
AK159670.1 - 4 - 5 789 75
AY026239.1 - 4 - 5 841 75
BC017621.1 - 4 - 5 1093 75
BC026639.1 - 4 - 5 782 75

View Full Product Details