The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007644.3 | NP_031670.1 | 2 - 3 | 572 | 134 |
GenBank mRNA | AB008553.1 | - | 2 - 3 | 477 | 134 |
AK011123.1 | - | 2 - 3 | 502 | 134 | |
AK083038.1 | - | 2 - 3 | 573 | 134 | |
AK144235.1 | - | 2 - 3 | 604 | 134 | |
AK149102.1 | - | - | 518 | 134 | |
BC029073.1 | - | 2 - 3 | 480 | 134 |