The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013648.6 | NP_038676.1 | 1 - 2 | 244 | 78 |
GenBank mRNA | AF038538.1 | - | 1 - 2 | 200 | 78 |
AK144271.1 | - | 1 - 2 | 207 | 78 | |
AK165915.1 | - | 1 - 2 | 234 | 78 | |
AU066588.1 | - | 1 - 2 | 245 | 78 | |
BC031370.1 | - | 1 - 2 | 201 | 78 | |
CJ176292.1 | - | 1 - 2 | 211 | 78 |