The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021451.2 | NP_067426.1 | 2 - 3 | 365 | 82 |
GenBank mRNA | AB041230.1 | - | 2 - 3 | 365 | 82 |
AK043856.1 | - | 2 - 3 | 396 | 82 | |
AK088556.1 | - | 2 - 3 | 375 | 82 | |
AK143990.1 | - | 2 - 3 | 437 | 82 | |
AK169914.1 | - | 2 - 3 | 409 | 82 | |
BC050821.1 | - | 2 - 3 | 352 | 82 |