The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167705.1 | NP_001161177.1 | 4 - 5 | 1287 | 79 |
NM_028740.2 | NP_083016.1 | 3 - 4 | 627 | 79 | |
GenBank mRNA | AK016702.1 | - | 3 - 4 | 630 | 79 |
BC063325.1 | - | 4 - 5 | 1477 | 79 |