The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139307.3 | NP_647468.2 | 1 - 2 | 200 | 59 |
GenBank mRNA | AJ458938.1 | - | 1 - 2 | 141 | 59 |
AK012169.1 | - | 1 - 2 | 173 | 59 | |
AK083684.1 | - | 1 - 2 | 186 | 59 | |
AK132325.1 | - | 1 - 2 | 205 | 59 | |
BC050274.1 | - | 1 - 2 | 151 | 59 |