Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oit1
Entrez Gene ID
18300
Gene Name
oncoprotein induced transcript 1
Gene Aliases
2310076N21Rik, AV067083, EF-7, Fam3d
Chromosome Location
Chr.14: 8348937 - 8378763 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146050.2 NP_666162.1 2 - 3 259 84
GenBank mRNA AK009397.1 - 2 - 3 234 84
AK010206.1 - 2 - 3 234 84
BC024504.1 - 2 - 3 242 84
BC024532.1 - 2 - 3 75 84
BC024542.1 - 2 - 3 256 84
BC034193.1 - 2 - 3 265 84

View Full Product Details