The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146126.4 | NP_666238.1 | 9 - 9 | 1835 | 91 |
GenBank mRNA | AK004692.1 | - | 9 - 9 | 1859 | 91 |
AK015059.1 | - | 9 - 9 | 1836 | 91 | |
AK152169.1 | - | 9 - 9 | 1884 | 91 | |
AK166988.1 | - | 9 - 9 | 1835 | 91 | |
BC024124.1 | - | 9 - 9 | 1850 | 91 | |
BC030875.1 | - | 9 - 9 | 1824 | 91 | |
BC039957.1 | - | 9 - 9 | 1835 | 91 | |
BC092291.1 | - | 9 - 9 | 1838 | 91 |