Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H2-T9
- Entrez Gene ID
-
15051
- Gene Name
- histocompatibility 2, T region locus 9
- Gene Aliases
-
H-2T9
- Chromosome Location
-
-
- Gene Symbol
- H2-T10
- Entrez Gene ID
-
15024
- Gene Name
- histocompatibility 2, T region locus 10
- Gene Aliases
-
H-2T10
- Chromosome Location
-
Chr.17: 36115871 - 36121444 on Build GRCm38
- Gene Symbol
- H2-T22
- Entrez Gene ID
-
15039
- Gene Name
- histocompatibility 2, T region locus 22
- Gene Aliases
-
27b, H-2T17, H-2T22, H2-T17, T22b
- Chromosome Location
-
Chr.17: 36037133 - 36042742 on Build GRCm38
View Full Product Details