Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Orc5
Entrez Gene ID
26429
Gene Name
origin recognition complex, subunit 5
Gene Aliases
AL033327, MmORC5, Orc5l
Chromosome Location
Chr.5: 22486484 - 22550367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011959.2 NP_036089.1 7 - 8 801 93
GenBank mRNA AJ007360.1 - 7 - 8 798 93
AK084683.1 - 7 - 8 829 93
AK144983.1 - 7 - 8 817 93
AK165912.1 - 7 - 8 807 93
AK166537.1 - 7 - 8 740 93
BC006927.1 - 7 - 8 817 93

View Full Product Details