Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Akr7a5
Entrez Gene ID
110198
Gene Name
aldo-keto reductase family 7, member A5 (aflatoxin aldehyde reductase)
Gene Aliases
0610025K21Rik, Afar, Afar1, Akr7a2
Chromosome Location
Chr.4: 139310738 - 139318786 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025337.3 NP_079613.3 2 - 3 526 87
GenBank mRNA AF525358.2 - 2 - 3 428 87
AJ271800.1 - 2 - 3 443 87
AJ271801.1 - 2 - 3 443 87
AK003915.1 - 2 - 3 513 87
AK143203.1 - 2 - 3 525 87
BC031857.1 - 2 - 3 503 87

View Full Product Details