The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199349.1 | NP_001186278.1 | 1 - 2 | 324 | 82 |
NM_138309.3 | NP_612182.1 | 1 - 2 | 324 | 82 | |
GenBank mRNA | AK031264.1 | - | 1 - 2 | 81 | 82 |
AK054364.1 | - | 1 - 2 | 124 | 82 | |
AK080225.1 | - | 1 - 2 | 80 | 82 | |
AK166275.1 | - | 1 - 2 | 75 | 82 | |
AY078163.1 | - | 1 - 2 | 120 | 82 | |
BC031736.1 | - | 1 - 2 | 99 | 82 | |
BY727540.1 | - | 1 - 2 | 325 | 82 | |
EF516990.1 | - | 1 - 2 | 82 | 82 |