The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114665.2 | NP_001108137.1 | 15 - 16 | 1811 | 71 |
NM_153118.3 | NP_694758.2 | 13 - 14 | 1637 | 71 | |
GenBank mRNA | BC034510.1 | - | 8 - 9 | 1085 | 71 |