Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ano2
Entrez Gene ID
243634
Gene Name
anoctamin 2
Gene Aliases
BC033409, Tmem16b
Chromosome Location
Chr.6: 125690387 - 126040133 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153589.2 NP_705817.2 17 - 18 2132 64
GenBank mRNA AK132944.1 - 9 - 10 930 64
BC033409.1 - 16 - 17 1727 64
BC060368.1 - 17 - 18 1821 64
FJ384096.1 - 17 - 18 2133 64
KC164759.2 - 16 - 17 2120 64
KC164760.2 - 15 - 16 2021 64
KC164761.2 - 15 - 16 1813 64
KC164762.2 - 14 - 15 1714 64

View Full Product Details