The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252374.1 | NP_001239303.1 | 8 - 9 | 658 | 125 |
NM_026004.3 | NP_080280.3 | 7 - 8 | 815 | 125 | |
GenBank mRNA | AK011525.1 | - | 7 - 8 | 766 | 125 |
AK011894.1 | - | 7 - 8 | 713 | 125 | |
BC038029.1 | - | 8 - 9 | 651 | 125 |