Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtpbp4
Entrez Gene ID
69237
Gene Name
GTP binding protein 4
Gene Aliases
2610028C09Rik, Crfg, Gtpbp3, NGB, Nog1
Chromosome Location
Chr.13: 8972464 - 8996012 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027000.4 NP_081276.2 5 - 6 576 67
GenBank mRNA AF325354.1 - 5 - 6 560 67
AF348208.1 - 5 - 6 583 67
AK011580.1 - 5 - 6 587 67
AK141852.1 - 5 - 6 586 67
AK163372.1 - 4 - 5 383 67
AK164330.1 - 5 - 6 577 67
AK166711.1 - 5 - 6 582 67
BC005514.1 - 5 - 6 557 67
BC025431.1 - 5 - 6 581 67

View Full Product Details