The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145619.2 | NP_663594.2 | 2 - 3 | 339 | 79 |
GenBank mRNA | AF368233.1 | - | 2 - 3 | 339 | 79 |
AK044223.1 | - | 2 - 3 | 317 | 79 | |
AK145259.1 | - | 2 - 3 | 358 | 79 | |
AK147868.1 | - | 2 - 3 | 638 | 79 | |
AK165070.1 | - | 2 - 3 | 269 | 79 | |
AK170541.1 | - | 2 - 3 | 283 | 79 | |
BC014870.1 | - | 2 - 3 | 279 | 79 | |
BC058754.1 | - | 2 - 3 | 259 | 79 | |
BC062204.1 | - | 2 - 3 | 259 | 79 |